missing translation for 'onlineSavingsMsg'
Learn More

VTX-27, MedChemExpress

Product Code. 16420997
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16420997 50 mg 50mg
16495526 100 mg 100mg
16460146 2 mg 2mg
16457417 5 mg 5mg
16443206 25 mg 25mg
16414708 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16420997 Supplier Medchem Express Supplier No. HY112782

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VTX-27 is a selective protein kinase C θ (PKC θ) inhibitor, with Kis of 0.08 nM and 16 nM for PKC θ and PKC δ.

VTX-27 is a selective protein kinase C θ (PKC θ) inhibitor, with Kis of 0.08 nM and 16 nM for PKC θ and PKC δ.

Specifications

Chemical Name or Material VTX-27
CAS 1321924-70-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H24ClFN6O
Quantity 50 mg
Solubility Information DMSO : 125 mg/mL (298.40 mM; Need ultrasonic)
SMILES FC1=C(N=C(N2C[C@@](NCC2)([H])[C@](C)(O)C(C)C)C(Cl)=C1)C3=NNC4=NC=CC=C34
Molecular Weight (g/mol) 418.9
Formula Weight 418.9
Percent Purity 97.01%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.