missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VTI1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116120
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total VTI1B.
V-SNARE that mediates vesicle transport pathways through interactions with t-SNAREs on the target membrane. These interactions are proposed to mediate aspects of the specificity of vesicle trafficking and to promote fusion of the lipid bilayers. May be concerned with increased secretion of cytokines associated with cellular senescence. Fischer von Mollard G., J. Biol. Chem. 273:2624-2630(1998). Li H.-C., Biochem. Biophys. Res. Commun. 247:70-74(1998).
Specifications
| VTI1B | |
| Polyclonal | |
| Unconjugated | |
| Vti1b | |
| A novel SNARE; AU015348; GES30; HGNC:17793; MVti1b; SNARE; Soluble N-ethylmaleimide-sensitive factor-attachment protein receptor; vesicle transport through interaction with t-SNAREs 1B; vesicle transport through interaction with t-SNAREs 1B homolog; vesicle transport through interaction with t-SNAREs homolog 1B; Vesicle transport v-SNARE protein Vti1-like 1; vesicle-associated soluble NSF attachment protein receptor; v-SNARE; VTI1; VTI1B; VTI1L; VTI1L1; VTI1-LIKE; vti1-rp1; VTI2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 100359512, 10490, 53612 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O88384, P58200, Q9UEU0 | |
| Vti1b | |
| A synthesized peptide derived from human VTI1B(Accession Q9UEU0), corresponding to amino acid residues V117-L167. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction