missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VTI1B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | VTI1B |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Regulatory Status | RUO |
Description
VTI1B Polyclonal specifically detects VTI1B in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
| VTI1B | |
| Unconjugated | |
| RUO | |
| vesicle transport through interaction with t-SNAREs homolog 1B (yeast), Vesicle transport v-SNARE protein Vti1-like 1, vesicle-associated soluble NSF attachment protein receptor, v-SNARE, VTI1, VTI1L, VTI1L1, VTI1-LIKE, Vti1-rp1, VTI2vesicle transport through interaction with t-SNAREs homolog 1B | |
| VTI1B | |
| IgG | |
| 26 kDa |
| Polyclonal | |
| Rabbit | |
| NP_006361 | |
| 10490 | |
| The immunogen for this antibody is VTI1B. Peptide sequence IERSHRIATETDQIGSEIIEELGEQRDQLERTKSRLVNTSENLSKSRKIL. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title