missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VTI1B Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-96993V
This item is not returnable.
View return policy
Description
VTI1B Polyclonal specifically detects VTI1B in Human samples. It is validated for Western Blot.
Specifications
| VTI1B | |
| Polyclonal | |
| Western Blot | |
| vesicle transport through interaction with t-SNAREs homolog 1B (yeast), Vesicle transport v-SNARE protein Vti1-like 1, vesicle-associated soluble NSF attachment protein receptor, v-SNARE, VTI1, VTI1L, VTI1L1, VTI1-LIKE, Vti1-rp1, VTI2vesicle transport through interaction with t-SNAREs homolog 1B | |
| Produced in rabbits immunized with purified, recombinant VTI1B (NP_006361.1; Ala 2-Leu 208). | |
| 0.1 mL | |
| Signal Transduction | |
| 10490 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction