missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VT 104

Product Code. 30713261
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30713261 50 mg 50mg
30713260 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30713261 Supplier Tocris Bioscience™ Supplier No. 8176/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent pan-TEAD inhibitor

VT 104 is a potent pan-TEAD (transcriptional enhanced associate domain) inhibitor (IC50 = 10.4 nM). VT 104 inhibits YAP/TAZ-TEAD promoted gene transcription by preventing the association between YAP/TAZ and TEAD. VT 104 blocks TEAD auto-palmitoylation. In vitro, VT 104 inhibits NF2-deficient mesothelioma cell proliferation, and growth of subcutaneous tumor xenografts in vivo. VT 104 is orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[(1S)-1-(2-Pyridinyl)ethyl]-5-[4-(trifluoromethyl)phenyl]-2-naphthalenecarboxamide
CAS 2417718-25-1
Quantity 50 mg
Target YAP1
Molecular Formula C25H19F3N2O
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.