missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VT 104
Description
VT 104 is a potent pan-TEAD (transcriptional enhanced associate domain) inhibitor (IC50 = 10.4 nM). VT 104 inhibits YAP/TAZ-TEAD promoted gene transcription by preventing the association between YAP/TAZ and TEAD. VT 104 blocks TEAD auto-palmitoylation. In vitro, VT 104 inhibits NF2-deficient mesothelioma cell proliferation, and growth of subcutaneous tumor xenografts in vivo. VT 104 is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | N-[(1S)-1-(2-Pyridinyl)ethyl]-5-[4-(trifluoromethyl)phenyl]-2-naphthalenecarboxamide |
| CAS | 2417718-25-1 |
| Quantity | 10 mg |
| Target | YAP1 |
| Molecular Formula | C25H19F3N2O |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?