missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSX1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23566-1-AP-20UL
This item is not returnable.
View return policy
Description
VSX1 contains a paired-like homeodomain and binds to the core of the locus control region of the red/green visual pigment gene cluster. VSX1 may regulate expression of the cone opsin genes early in development. Mutations in this gene can cause posterior polymorphous corneal dystrophy (PPCD) and keratoconus. The protein encoded by this gene contains a paired-like homeodomain and binds to the core of the locus control region of the red/green visual pigment gene cluster. The encoded protein may regulate expression of the cone opsin genes early in development. Mutations in this gene can cause posterior polymorphous corneal dystrophy (PPCD) and keratoconus. Two transcript variants encoding different isoforms have been found for this gene. The protein encoded by this gene contains a paired-like homeodomain and binds to the core of the locus control region of the red/green visual pigment gene cluster. The encoded protein may regulate expression of the cone opsin genes early in development. Mutations in this gene can cause posterior polymorphous corneal dystrophy and keratoconus. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
| VSX1 | |
| Polyclonal | |
| Unconjugated | |
| VSX1 | |
| Homeodomain protein RINX, KTCN, PPCD, PPD, RINX, Transcription factor VSX1, visual system homeobox 1, VSX1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114889, 30813 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q91V10, Q9NZR4 | |
| Vsx1 | |
| VSX1 Fusion Protein Ag19222 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu