missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSX1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £358.00
Specifications
| Antigen | VSX1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
VSX1 Polyclonal antibody specifically detects VSX1 in Human samples. It is validated for Western BlotSpecifications
| VSX1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Neuroscience | |
| PBS with 50% glycerol, pH7.3. | |
| 30813 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CAASDS, KTCN, KTCN1, PPCD, PPD, RINX, VSX1 visual system homeobox 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human VSX1 (NP_055403.2). MTGRDSLSDGRTSSRALVPGGSPRGSRPRGFAITDLLGLEAELPAPAGPGQGSGCEGPAVAPCPGPGLDGSSLARGALPLGLGLLCGFGTQPPAAARAPCLLLADVPFLPPRGPEPAAPLAPSRPPPALGRQKRSDSVSTSDEDSQSEDRNDLKASPTLGKRKKRRHRTV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title