missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VSTM2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548140
This item is not returnable.
View return policy
Description
VSTM2A Polyclonal Antibody for Western Blot, IHC (P)
Plays a role in the regulation of the early stage of white and brown preadipocyte cell differentiation. Promotes adipogenic commitment of preadipocytes by increasing gene expression of the transcription factor PPARG in a BMP4-dependent signaling pathway.
Specifications
| VSTM2A | |
| Polyclonal | |
| Unconjugated | |
| VSTM2A | |
| hypothetical protein MGC33530; V-set and transmembrane domain containing 2; V-set and transmembrane domain containing 2A; V-set and transmembrane domain-containing protein 2A; Vstm2; VSTM2A | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 211739, 222008 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8R0A6, Q8TAG5 | |
| VSTM2A | |
| KLH conjugated synthetic peptide between 97-126 amino acids from the Central region of human VSTM2A. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction