missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSTM2A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£231.00 - £571.00
Specifications
| Antigen | VSTM2A |
|---|---|
| Dilution | Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:1000-1:2500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18450291
|
Novus Biologicals
NBP1-81113-25ul |
25ul |
£231.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18247535
|
Novus Biologicals
NBP1-81113 |
0.1 mL |
£571.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VSTM2A Polyclonal specifically detects VSTM2A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| VSTM2A | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 222008 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KFTEFPRNVTATEGQNVEMSCAFQSGSASVYLEIQWWFLRGPEDLDPGAEGAGAQVELLPDRDPDSDGTKISTVK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:1000-1:2500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| MGC33530, V-set and transmembrane domain containing 2, V-set and transmembrane domain containing 2A, V-set and transmembrane domain-containing protein 2A, VSTM2 | |
| VSTM2A | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title