missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSTM1 Antibody (104), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90190B
This item is not returnable.
View return policy
Description
VSTM1 Monoclonal specifically detects VSTM1 in Human samples. It is validated for ELISA.
Specifications
| VSTM1 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 284415 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 104 | |
| ELISA | |
| LAIR homolog, MGC119160, MGC119161, OSCAR-like transcript-1, signal inhibitory receptor on leukocytes-1, SIRL1, UNQ3033, V-set and transmembrane domain containing 1, V-set and transmembrane domain-containing protein 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human VSTM1 (Q6UX27-16; Met1-Thr135). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction