missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSP15a Rabbit anti-Yeast, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | VSP15a |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16816385
|
Proteintech
51090-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16806385
|
Proteintech
51090-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody recognizes the SPAPB1A10 protein of Yeast.
Specifications
| VSP15a | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Yeast | |
| SPAPB1A10, VSP15a | |
| SPAPB1A10.05 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| 2543600 | |
| VSP15a Fusion Protein Ag0724 | |
| Primary | |
| -20°C | |
| SPAPB1A10.05 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title