missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VSNL1 Polyclonal Antibody

Chicken Polyclonal Antibody
Brand: Invitrogen™ PA5143584
This item is not returnable.
View return policy
Description
VSNL1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IHC (PFA)
This gene is a member of the visinin/recoverin subfamily of neuronal calcium sensor proteins. The encoded protein is strongly expressed in granule cells of the cerebellum where it associates with membranes in a calcium-dependent manner and modulates intracellular signaling pathways of the central nervous system by directly or indirectly regulating the activity of adenylyl cyclase. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined.
Specifications
| VSNL1 | |
| Polyclonal | |
| Unconjugated | |
| VSNL1 | |
| 21 kDa CABP; hippocalcin-like protein 3; HLP3; HPCAL3; HUVISL1; Neural visinin-like protein 1; neurocalcin alpha; Neurocalcin-alpha; NVL-1; Nvp1; NVP-1; Ratnvp1; VILIP; VILIP-1; visinin like 1; visinin-like 1; visinin-like protein 1; Visl1; VLP-1; Vnsl1; VSNL1 | |
| Chicken | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Bovine, Pig | |
| Antibody | |
| IgY |
| Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| Conc. Not Determined | |
| PBS with 0.02% sodium azide | |
| P62760, P62761, P62762, P62763 | |
| VSNL1 | |
| Full length recombinant human VLP1. | |
| RUO | |
| 100517707, 24877, 26950, 282124, 7447 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction