missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VSNL1 Monoclonal Antibody (3A9)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA547408

 View more versions of this product

Product Code. 17896847

  • £430.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

VSNL1 Monoclonal Antibody for Western Blot, ICC/IF, IHC

This gene is a member of the visinin/recoverin subfamily of neuronal calcium sensor proteins. The encoded protein is strongly expressed in granule cells of the cerebellum where it associates with membranes in a calcium-dependent manner and modulates intracellular signaling pathways of the central nervous system by directly or indirectly regulating the activity of adenylyl cyclase. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VSNL1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 5mM sodium azide
P62760, P62761, P62762, P62763
VSNL1
Recombinant full length human protein.
100 μL
Primary
Human, Mouse, Rat, Bovine, Pig
Antibody
IgG1
Immunohistochemistry, Western Blot, Immunocytochemistry
3A9
Unconjugated
VSNL1
21 kDa CABP; hippocalcin-like protein 3; HLP3; HPCAL3; HUVISL1; Neural visinin-like protein 1; neurocalcin alpha; Neurocalcin-alpha; NVL-1; Nvp1; NVP-1; Ratnvp1; VILIP; VILIP-1; visinin like 1; visinin-like 1; visinin-like protein 1; Visl1; VLP-1; Vnsl1; VSNL1
Mouse
Protein A/G
RUO
100517707, 24877, 26950, 282124, 7447
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.