missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSIR, Mouse anti-Human, Clone: CL3975, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB15698.100uL
This item is not returnable.
View return policy
Description
Sequence: LTFQDLHLHHGGHQAANTSHDLAQRHGLESASDHHGNFSITMRNLTLLDSGLYCCLVVEIRHHHSEHRVHGAMELQVQTGKDAPSNCVVYPSSSSpecifications
| VSIR | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
| B7-H5/B7H5/C10orf54/DD1alpha/GI24/PD-1H/PP2135/SISP1/VISTA | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| CL3975 | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:500) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| V-set immunoregulatory receptor | |
| VSIR | |
| Recombinant protein corresponding to human VSIR. | |
| RUO | |
| 64115 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction