missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VSIG8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551157
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total VSIG8 protein.
VSIG8 contains 2 Ig-like V-type (immunoglobulin-like) domains. VSIG8 is single-pass type I membrane protein. The function of the VSIG8 protein remains unknown.
Specifications
| VSIG8 | |
| Polyclonal | |
| Unconjugated | |
| VSIG8 | |
| A030011M19; C1orf204; EG240916; LOC100727929; LOW QUALITY PROTEIN: V-set and immunoglobulin domain-containing protein 8; RGD1562464; V-set and immunoglobulin domain containing 8; V-set and immunoglobulin domain containing 8; V-set and immunoglobulin domain-containing protein 8; V-set and immunoglobulin domain-containing protein 8; Vsig8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 240916, 391123 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.7 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P0DPA2, Q6P3A4 | |
| VSIG8 | |
| Fusion protein of human VSIG8. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction