missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSIG4 Monoclonal Antibody (03)

VSIG4 Monoclonal Antibody (3)
Brand: Invitrogen MA531001
This item is not returnable.
View return policy
Description
Specifications
| VSIG4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9Y279 | |
| VSIG4 | |
| Recombinant Human VSIG4 protein (Met1-Pro283). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA | |
| 03 | |
| Unconjugated | |
| VSIG4 | |
| A530061A11; BC025105; complement receptor of the immunoglobulin superfamily; CRIg; Ig superfamily protein; MGC137243 protein; protein Z39Ig; UNQ317/PRO362; V-set and immunoglobulin domain containing 4; V-set and immunoglobulin domain-containing protein 4; VSIG4; Z39IG | |
| Mouse | |
| Protein A | |
| RUO | |
| 11326 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction