missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSIG4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£285.00 - £451.00
Specifications
| Antigen | VSIG4 |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200-1:500 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18414001
|
Novus Biologicals
NBP1-86843-25ul |
25 μL |
£285.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18717413
|
Novus Biologicals
NBP1-86843 |
0.1 mL |
£451.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VSIG4 Polyclonal specifically detects VSIG4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| VSIG4 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q9Y279 | |
| 11326 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:PPISYIWYKQQTNNQEPIKVATLSTLLFKPAVIADSGSYFCTAKGQVGSEQHSDIVKFVVKDSSKLLKTKTEAPTTMTYPLKATSTVKQSWDWTTDMDGYL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 44 kDa |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200-1:500 | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IGcomplement receptor of the immunoglobulin superfamily | |
| VSIG4 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title