missing translation for 'onlineSavingsMsg'
Learn More

VSIG3/IGSF11 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18353497 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18353497 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18353497 Supplier Novus Biologicals Supplier No. NBP300068AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VSIG3/IGSF11 Polyclonal specifically detects VSIG3/IGSF11 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VSIG3/IGSF11
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias Brain and testis-specific immunoglobulin superfamily protein, Bt-IGSF, BTIGSF, cancer/testis antigen 119, CT119, CXADR like 1, CXADRL1, IgSF11, Igsf13, immunoglobulin superfamily member 11, immunoglobulin superfamily, member 11, MGC35227, V-set and immunoglobulin domain containing 3, V-set and immunoglobulin domain-containing protein 3, VSIG3brain and testis-specific immunoglobin superfamily protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant VSIG3/IGSF11 (AAH34411.1; Met1-Gly240).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152404
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.