missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSIG2 Antibody (541522), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB51771MFV500
This item is not returnable.
View return policy
Description
VSIG2 Monoclonal antibody specifically detects VSIG2 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| VSIG2 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 541522 | |
| Flow Cytometry, CyTOF-ready | |
| 2210413P10Rik, Cortical thymocyte-like protein, CTHCTXLcortical thymocyte receptor (X. laevis CTX) like, CT-like protein, V-set and immunoglobulin domain containing 2, V-set and immunoglobulin domain-containing protein 2 | |
| Mouse myeloma cell line NS0-derived recombinant human VSIG2, Val24-Ala243, Accession # Q961Q7-1 | |
| RUO | |
| 23584 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction