missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VSIG10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571389
This item is not returnable.
View return policy
Description
VSIG10 Polyclonal Antibody for Western Blot
VSIG10 (V-Set And Immunoglobulin Domain Containing 10) is a Protein Coding gene. Diseases associated with VSIG10 include Adenoid Hypertrophy. An important paralog of this gene is CNTN5. [GeneCards].
Specifications
| VSIG10 | |
| Polyclonal | |
| Unconjugated | |
| VSIG10 | |
| RGD1565800; V-set and immunoglobulin domain containing 10; V-set and immunoglobulin domain-containing protein 10; Vsig10 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54621 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N0Z9 | |
| VSIG10 | |
| synthetic peptide directed towards the C terminal region of human VSIG10. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction