missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VSIG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551580
This item is not returnable.
View return policy
Description
Immunogen sequence: QAVAIGQFKD RITGSNDPGN ASITISHMQP ADSGIYICDV NNPPDFLGQN QGILNVSVLV KPSKPLCSVQ GRPETGHTIS LSCLSALGTP SPVYYWHKLE GRDIVPV Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 82%.
VSIG1 encodes a member of the junctional adhesion molecule (JAM) family. The encoded protein contains multiple glycosylation sites at the N-terminal region, and multiple phosphorylation sites and glutamic acid/proline (EP) repeats at the C-terminal region. The gene is expressed in normal stomach and testis, as well as in gastric, esophageal and ovarian cancers. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| VSIG1 | |
| Polyclonal | |
| Unconjugated | |
| VSIG1 | |
| 1700062D20Rik; 4930405J24Rik; cell surface A33 antigen; CHT1; ChT1 thymocyte antigen; dJ889N15.1; Glycoprotein A34; GPA34; MGC44287; V-set and immunoglobulin domain containing 1; V-set and immunoglobulin domain-containing protein 1; Vsig1; VSIG1 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 340547 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86XK7 | |
| VSIG1 | |
| Recombinant protein corresponding to Human VSIG1. Recombinant protein control fragment (Product #RP-91992). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction