Learn More
Invitrogen™ VSIG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589497
Description
Immunogen sequence: ASREQLSIQW SFFHKKEMEP ISIYFSQGGQ AVAIGQFKDR ITGSNDPGNA SITISHMQPA DSGIYICDVN NPPDFLGQNQ GILNVSVLVK PSKPLCSVQG RPETGHTISL SCLSALGTPS PVYYWHKLEG RDIVPVKENF NPTTGILVIG NLTNFEQGYY QCTAINRLGN SSCEIDLTSS H; Positive Samples: BxPC3, OVCAR3, Mouse testis, Mouse stomach, Mouse pancreas; Cellular Location: Membrane, Single-pass type I membrane protein.
VSIG1 encodes a member of the junctional adhesion molecule (JAM) family. The encoded protein contains multiple glycosylation sites at the N-terminal region, and multiple phosphorylation sites and glutamic acid/proline (EP) repeats at the C-terminal region. The gene is expressed in normal stomach and testis, as well as in gastric, esophageal and ovarian cancers. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| VSIG1 | |
| Polyclonal | |
| Unconjugated | |
| VSIG1 | |
| 1700062D20Rik; 4930405J24Rik; cell surface A33 antigen; CHT1; ChT1 thymocyte antigen; dJ889N15.1; Glycoprotein A34; GPA34; MGC44287; V-set and immunoglobulin domain containing 1; V-set and immunoglobulin domain-containing protein 1; Vsig1; VSIG1 protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 315920, 340547, 78789 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.46 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q4KLY3, Q86XK7, Q9D2J4 | |
| VSIG1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 50-230 of human VSIG1 (NP_8724131). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.