missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VSIG1 Monoclonal Antibody (CT1), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528800
This item is not returnable.
View return policy
Description
VSIG1 Monoclonal Antibody for Flow
VSIG1 encodes a member of the junctional adhesion molecule (JAM) family. The encoded protein contains multiple glycosylation sites at the N-terminal region, and multiple phosphorylation sites and glutamic acid/proline (EP) repeats at the C-terminal region. The gene is expressed in normal stomach and testis, as well as in gastric, esophageal and ovarian cancers. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| VSIG1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with proprietary stabilizer and <0.1% sodium azide; pH 7.4 | |
| Q9PWR4 | |
| VSIG1 | |
| Chicken thymocytes. | |
| 1 mL | |
| Primary | |
| Avian, Chicken | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| CT1 | |
| FITC | |
| VSIG1 | |
| 1700062D20Rik; 4930405J24Rik; cell surface A33 antigen; CHT1; ChT1 thymocyte antigen; dJ889N15.1; Glycoprotein A34; GPA34; MGC44287; V-set and immunoglobulin domain containing 1; V-set and immunoglobulin domain-containing protein 1; Vsig1; VSIG1 protein | |
| Mouse | |
| Purified | |
| RUO | |
| 414795 | |
| 4°C, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction