missing translation for 'onlineSavingsMsg'
Learn More

VS-5584, MedChemExpress

Product Code. 16493127
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493127 50 mg 50mg
16467088 10 mg 10mg
16419637 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16493127 Supplier Medchem Express Supplier No. HY16585

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress VS-5584 is a pan-PI3K/mTOR kinase inhibitor with IC50s of 16 nM, 68 nM, 42 nM, 25 nM, and 37 nM for PI3Kα, PI3Kβ, PI3Kδ, PI3Kγ and mTOR, respectively. VS-5584 simultaneously blocks mTORC2 as well as mTORC1.

VS-5584 is a pan-PI3K/mTOR kinase inhibitor with IC50s of 16 nM, 68 nM, 42 nM, 25 nM, and 37 nM for PI3Kα, PI3Kβ, PI3Kδ, PI3Kγ and mTOR, respectively. VS-5584 simultaneously blocks mTORC2 as well as mTORC1.

Specifikationer

Chemical Name or Material VS-5584
CAS 1246560-33-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H22N8O
Quantity 50 mg
Synonym SB2343
Solubility Information DMSO : 33.33 mg/mL (94.04 mM; Need ultrasonic)
SMILES NC1=NC=C(C2=C3N=C(C)N(C(C)C)C3=NC(N4CCOCC4)=N2)C=N1
Molecular Weight (g/mol) 354.41
Formula Weight 354.41
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Visa mer Visa mindre

Research purposes only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.