missing translation for 'onlineSavingsMsg'
Learn More

VRL1 Antibody - BSA Free, Novus Biologicals™

Product Code. 18408475 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.01mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408475 0.05 mg 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408475 Supplier Novus Biologicals Supplier No. NBP225068

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VRL1 Polyclonal antibody specifically detects VRL1 in Human, Porcine, Equine, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen VRL1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 10 μg/mL
Formulation PBS
Gene Accession No. Q9Y5S1
Gene Alias Osm-9-like TRP channel 2, transient receptor potential cation channel, subfamily V, member 2, TrpV2, Vanilloid receptor-like protein 1, VRL-1MGC12549, VRL1transient receptor potential cation channel subfamily V member 2, VRLOTRPC2
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from N-terminus of human VRL1.
Purification Method Immunogen affinity purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51393
Target Species Human, Pig, Equine, Monkey
Content And Storage Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.