missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VRK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12946-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the vaccinia-related kinase family of serine/threonine protein kinases. This gene is widely expressed in human tissues and has increased expression in actively dividing cells, such as those in testis, leukocytes, fetal liver, and carcinomas. Its protein localizes to the endoplasmic reticulum and has been shown to phosphorylate casein and undergo autophosphorylation. Multiple alternatively spliced transcript variants have been found for this gene.Specifications
| VRK2 | |
| Polyclonal | |
| Unconjugated | |
| VRK2 | |
| vaccinia related kinase 2, VRK2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 360991, 69922, 7444 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q86Y07, Q8BN21 | |
| Vrk2 | |
| VRK2 Fusion Protein Ag4019 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction