missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VRK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101871
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total VRK2.
VRK2 serine/threonine kinases vaccinia related kinase 2 is a member of the vaccinia-related kinase (VRK) family of kinases. VRK2 is widely expressed in human tissues and highly expressed in actively dividing cells, such as those in testis, leukocytes, fetal liver and carcinomas. VRK2 can be used to phosphorylate casein and itself undergoes autophosphorylation. VRK2 interacts specifically with Epstein-Barr virus BHRF1, a homologue of Bcl-2, and this interaction is involved in protecting cells from apoptosis.
Specifications
| VRK2 | |
| Polyclonal | |
| Unconjugated | |
| VRK2 | |
| 2810003O05Rik; AI447698; Serine/threonine-protein kinase VRK2; vaccinia related kinase 2; vaccinia virus B1R-related kinase 2; vaccinia virus BIR kinase related kinase 2; vaccinia-related kinase 2; Vrk2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 69922, 7444 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q86Y07, Q8BN21 | |
| VRK2 | |
| A synthesized peptide derived from human VRK2(Accession Q86Y07), corresponding to amino acid residues G317-E367. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction