missing translation for 'onlineSavingsMsg'
Learn More

VRK1 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18379472 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379472 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379472 Supplier Novus Biologicals Supplier No. NBP299819R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VRK1 Polyclonal specifically detects VRK1 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VRK1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11.1, MGC117401, MGC138280, MGC142070, PCH1, serine/threonine-protein kinase VRK1, vaccinia related kinase 1, vaccinia virus B1R-related kinase 1, Vaccinia-related kinase 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant VRK1 (Q99986; Met1-Lys396).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Protein Kinase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7443
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.