Learn More
Grant Instruments™ VR Rack for Grant Heated Circulating Bath
For use with a number of Grant Heating Circulators and Heated Circulating Baths. Grant Instruments™ VR Racks for Grant Heated Circulating Baths accommodate a variety of tube sizes and capacities.
Brand:  Grant Instruments™ VR-LE
Code : 57
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11486477
  • £135.00 / Each
In Stock
to see stock.
Add to basket
Description
Description
For use with a number of Grant Heating Circulators and Heated Circulating Baths. Grant Instruments™ VR Racks for Grant Heated Circulating Baths accommodate a variety of tube sizes and capacities.
  • Accommodates tubes as small as 0.5mL or as large as 30mm dia.
  • For use with Grant Heated Circulating Baths and Heating Circulators.
Specifications
Specifications
T100, TC120, TX150, TXF200 Heating Circulators
(75) 1.5 mL Tubes