missing translation for 'onlineSavingsMsg'
Learn More

VPS8 Polyclonal Antibody
GREENER_CHOICE

VPS8 Polyclonal Antibody

Brand:  Thermo Scientific PA557855

Product Code. 15795741

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

VPS8 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY
Specifications

Specifications

VPS8
Polyclonal
Unconjugated
VPS8
AI315068; AU040738; im:7166005; KIAA0804; mKIAA0804; RGD1309443; si:dkey-24b15.1; vacuolar protein sorting 8 homolog; vacuolar protein sorting 8 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 8 homolog; VPS8; VPS8 CORVET complex subunit; VPS8 subunit of CORVET complex; VPS8, CORVET complex subunit; zgc:158695
Rabbit
Antigen Affinity Chromatography
RUO
23355
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8N3P4
VPS8
Recombinant protein corresponding to Human VPS8. Recombinant protein control fragment (Product #RP-96382).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.