missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPS8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569218
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Plays a role in vesicle-mediated protein trafficking of the endocytic membrane transport pathway. Believed to act as a component of the putative CORVET endosomal tethering complexes which is proposed to be involved in the Rab5-to-Rab7 endosome conversion probably implicating MON1A/B, and via binding SNAREs and SNARE complexes to mediate tethering and docking events during SNARE-mediated membrane fusion. The CORVET complex is proposed to function as a Rab5 effector to mediate early endosome fusion probably in specific endosome subpopulations (PubMed:25266290). Functions predominantly in APPL1-containing endosomes (PubMed:25266290). [UniProt].
Specifications
| VPS8 | |
| Polyclonal | |
| Unconjugated | |
| VPS8 | |
| AI315068; AU040738; im:7166005; KIAA0804; mKIAA0804; RGD1309443; si:dkey-24b15.1; vacuolar protein sorting 8 homolog; vacuolar protein sorting 8 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 8 homolog; VPS8; VPS8 CORVET complex subunit; VPS8 subunit of CORVET complex; VPS8, CORVET complex subunit; zgc:158695 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23355 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N3P4 | |
| VPS8 | |
| synthetic peptide directed towards the N-terminal region of human VPS8. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction