missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPS54 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571766
This item is not returnable.
View return policy
Description
VPS54 Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes for a protein that in yeast forms part of a trimeric vacuolar-protein-sorting complex that is required for retrograde transport of proteins from prevacuoles to the late Golgi compartment. As in yeast, mammalian Vps54 proteins contain a coiled-coil region and dileucine motifs. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| VPS54 | |
| Polyclonal | |
| Unconjugated | |
| Vps54 | |
| Hcc8; Hepatocellular carcinoma protein 8; hVps54L; mSLP8; PPP1R164; protein phosphatase 1, regulatory subunit 164; SLP-8p; Tumor antigen HOM-HCC-8; tumor antigen SLP-8p; Tumor antigen SLP-8p homolog; vacuolar protein sorting 54 (yeast); vacuolar protein sorting 54 homolog; vacuolar protein sorting 54 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 54; VPS54; VPS54 GARP complex subunit; VPS54, GARP complex subunit; Vps54l; vps54-like protein; Vsp54; wr | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 245944, 51542 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q5SPW0, Q9P1Q0 | |
| Vps54 | |
| KLH conjugated synthetic peptide between 465-491 amino acids from the Central region of human VPS54. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction