missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPS51 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521674
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, HeLa, HepG2. Predicted reactivity: Mouse (99%), Zebrafish (88%), Xenopus laevis (83%), Chicken (80%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Acts as component of the GARP complex that is involved in retrograde transport from early and late endosomes to the trans-Golgi network (TGN). The GARP complex is required for the maintenance of protein retrieval from endosomes to the TGN, acid hydrolase sorting, lysosome function, endosomal cholesterol traffic and autophagy. VPS51 participates in retrograde transport of acid hydrolase receptors, likely by promoting tethering and SNARE-dependent fusion of endosome-derived carriers to the TGN. Acts as component of the EARP complex that is involved in endocytic recycling. The EARP complex associates with Rab4-positive endosomes and promotes recycling of internalized transferrin receptor (TFRC) to the plasma membrane.
Specifications
| VPS51 | |
| Polyclonal | |
| Unconjugated | |
| VPS51 | |
| 1110014N23Rik; 3110057M17Rik; AI837850; ANG2; ANG3; Another new gene 2 protein; C11orf2; C11orf3; Ffr; PP5382; Protein fat-free homolog; vacuolar protein sorting 51 homolog; vacuolar protein sorting 51 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 51 homolog; VPS51; VPS51 GARP complex subunit; VPS51, GARP complex subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 738 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.26 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q9UID3 | |
| VPS51 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human C11orf2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction