missing translation for 'onlineSavingsMsg'
Learn More

VPS50 Antibody (2D11), Novus Biologicals™

Product Code. 18355709 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355709 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18355709 Supplier Novus Biologicals Supplier No. H00055610M01100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

VPS50 Monoclonal antibody specifically detects VPS50 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS50
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2D11
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_060137
Gene Alias CCDC132, coiled-coil domain containing 132, coiled-coil domain-containing protein 132, EARP/GARPII Complex Subunit VPS50, KIAA1861, Syndetin, VPS50, EARP/GARPII Complex Subunit, VPS54L
Host Species Mouse
Immunogen FLJ20097 (NP_060137, 862 a.a. ~ 964 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GYANVKKCSNEGRALMQLDFQQFLMKLEKLTDIRPIPDKEFVETYIKAYYLTENDMERWIKEHREYSTKQLTNLVNVCLGSHINKKARQKLLAAIDDIDRPKR
Purification Method IgG purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55610
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.