missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS4B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £484.00
Specifications
| Antigen | VPS4B |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18672819
|
Novus Biologicals
NBP2-49263-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18689276
|
Novus Biologicals
NBP2-49263 |
0.1 mL |
£484.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VPS4B Polyclonal antibody specifically detects VPS4B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| VPS4B | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Microautophagy | |
| PBS (pH 7.2), 40% Glycerol | |
| 9525 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Cell migration-inducing gene 1 protein, Protein SKD1, SKD1B, SKD1cell migration-inducing 1, Suppressor of K(+) transport growth defect 1, suppressor of K+ transport defect 1, vacuolar protein sorting 4 homolog B (S. cerevisiae), vacuolar protein sorting 4B, vacuolar protein sorting 4B (yeast), vacuolar protein sorting-associated protein 4B, VPS42, VPS4-2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: AQKPVKEGQPSPADEKGNDSDGEGESDDPE | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title