missing translation for 'onlineSavingsMsg'
Learn More

VPS4B Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18430545 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430545 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430545 Supplier Novus Biologicals Supplier No. NBP119118JF646

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VPS4B Polyclonal antibody specifically detects VPS4B in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS4B
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Cell migration-inducing gene 1 protein, Protein SKD1, SKD1B, SKD1cell migration-inducing 1, Suppressor of K(+) transport growth defect 1, suppressor of K+ transport defect 1, vacuolar protein sorting 4 homolog B (S. cerevisiae), vacuolar protein sorting 4B, vacuolar protein sorting 4B (yeast), vacuolar protein sorting-associated protein 4B, VPS42, VPS4-2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 300 and 350 of human vacuolar protein sorting 4 homolog B using the numbering given in entry NP_004860.2 (GeneID 9525).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Microautophagy
Primary or Secondary Primary
Gene ID (Entrez) 9525
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.