missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VPS4A Recombinant Rabbit Monoclonal Antibody (K01_2K09)
GREENER_CHOICE

Product Code. 30827137
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30827137 100 μL
30825813 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827137 Supplier Invitrogen™ Supplier No. MA563924

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

VPS4A Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

The protein encoded by this gene is a member of the AAA protein family (ATPases associated with diverse cellular activities), and is the homolog of the yeast Vps4 protein. In humans, two paralogs of the yeast protein have been identified. The former share a high degree of aa sequence similarity with each other, and also with yeast Vps4 and mouse Skd1 proteins. The mouse Skd1 (suppressor of K+ transport defect 1) has been shown to be really an yeast Vps4 ortholog. Functional studies indicate that both human paralogs associate with the endosomal compartments, and are involved in intracellular protein trafficking, similar to Vps4 protein in yeast. The gene encoding this paralog has been mapped to chromosome 16; the gene for the other resides on chromosome 18.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS4A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_2K09
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene VPS4A
Gene Accession No. Q793F9, Q8VEJ9, Q9UN37
Gene Alias 4930589C15Rik; AI325971; AW553189; hVPS4; Protein SKD2; SKD1; SKD1A; SKD1-homolog; SKD2; vacuolar protein sorting 4 homolog A; vacuolar protein sorting 4 homolog A (S. cerevisiae); vacuolar protein sorting 4a; vacuolar protein sorting 4a (yeast); vacuolar protein sorting factor 4A; vacuolar protein sorting protein 4a; vacuolar protein sorting-associated protein 4A; vacuolar sorting protein 4; VPS4; VPS4-1; VPS4A; vps4-A
Gene Symbols VPS4A
Host Species Rabbit
Immunogen A synthetic peptide of human VPS4a.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116733, 246772, 27183
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.