missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VPS41 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5142158

Product Code. 17856562

  • £406.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Additional Information: This antibody is expected to recognise both reported isoforms (NP_055211.2 and NP_542198.2). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.

Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human ortholog of yeast Vps41 protein which is also conserved in Drosophila, tomato, and Arabidopsis. Expression studies in yeast and human indicate that this protein may be involved in the formation and fusion of transport vesicles from the Golgi. Several transcript variants encoding different isoforms have been described for this gene, however, the full-length nature of not all is known.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VPS41
Polyclonal
Unconjugated
VPS41
AI317346; fa01f11; HVPS41; hVps41p; HVSP41; mVam2; RGD1560511; S53; si:ch211-146l10.2; vacuolar assembly protein 41; vacuolar protein sorting 41 (yeast); vacuolar protein sorting 41 homolog; vacuolar protein sorting 41 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 41 homolog; Vam2; VAM2 homolog; VPS41; VPS41 HOPS complex subunit; VPS41 subunit of HOPS complex; VPS41, HOPS complex subunit; wu:fa01f11
Goat
Ammonium Sulfate Precipitation
RUO
27072
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
P49754
VPS41
Peptide with sequence C-DHLKKDSQNKTLLK.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.