missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS39 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16219-1-AP-20UL
This item is not returnable.
View return policy
Description
VPS39 is the mammalian homolog of the yeast vacuolar protein VAM6, that in mammals, promotes lysosome clustering and fusion in vivo. VPS39 is also thought to modulate the TGF-beta response by coupling the TGF-beta receptor complex to the Smad pathway. Similar to the related protein TRAP1, the VPS39 protein is essential for early embryonic development, with mice lacking VPS39 dying at or before E6.5.Specifications
| VPS39 | |
| Polyclonal | |
| Unconjugated | |
| VPS39 | |
| hVam6p, KIAA0770, TLP, VAM6, Vam6/Vps39 like protein, VPS39 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23339, 269338 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8R5L3, Q96JC1 | |
| VPS39 | |
| VPS39 Fusion Protein Ag9267 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction