Learn More
VPS39 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE
Product Code. 16475064
Product Code. 16475064 Supplier Invitrogen Supplier No. PA588969
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Rabbit Polyclonal Antibody
Immunogen sequence: RFHNCLIQLY CEKVQGLMKE YLLSFPAGKT PVPAGEEEGE LGEYRQKLLM FLEISSYYDP GRLICDFPFD GLLEERALLL GRMGKHEQAL FIYVHILKDT RMAEEYCHKH YDRNKDGNKD VYLSLLRMYL SPPSIHCLGP IKLELLEPKA NLQAALQVLE LHHSKLDTTK ALNLLPANTQ INDIRIFLEK VLEENAQKKR FNQVLKNLLH AEFLRVQEER ILHQQVKCII TEEKVCMVCK KKIGNSAFAR YPNGVVVHYF CSKEVNPADT; Positive Samples: U-87MG, 293T, A-549; Cellular Location: Cytoplasm, Late endosome, Late endosome membrane, Lysosome, Lysosome membrane, Peripheral membrane protein
VPS39 is the mammalian homolog of the yeast vacuolar protein VAM6, that in mammals, promotes lysosome clustering and fusion in vivo. VPS39 is also thought to modulate the TGF-beta response by coupling the TGF-beta receptor complex to the Smad pathway. Similar to the related protein TRAP1, the VPS39 protein is essential for early embryonic development, with mice lacking VPS39 dying at or before E6. 5.