missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPS39 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100365
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total VPS39.
VPS39 is the mammalian homolog of the yeast vacuolar protein VAM6, that in mammals, promotes lysosome clustering and fusion in vivo. VPS39 is also thought to modulate the TGF-beta response by coupling the TGF-beta receptor complex to the Smad pathway. Similar to the related protein TRAP1, the VPS39 protein is essential for early embryonic development, with mice lacking VPS39 dying at or before E6. 5.
Specifications
| VPS39 | |
| Polyclonal | |
| Unconjugated | |
| VPS39 | |
| A230065P22Rik; AW743070; hVam6p; KIAA0770; mVam6; OTTMUSP00000023341; OTTMUSP00000023350; PI3K type 3; Pldn; TLP; TRAP1-like protein; vacuolar protein sorting 39 (yeast); vacuolar protein sorting 39 homolog; vacuolar protein sorting 39 homolog (S. cerevisiae); Vam6; Vam6/Vps39-like protein; Vam6P; Vps39; VPS39 HOPS complex subunit; VPS39, HOPS complex subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23339, 269338 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8R5L3, Q96JC1 | |
| VPS39 | |
| A synthesized peptide derived from human VPS39(Accession Q96JC1), corresponding to amino acid residues E410-I460. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction