missing translation for 'onlineSavingsMsg'
Learn More

VPS39 Antibody, Novus Biologicals™

Product Code. 18348958 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348958 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18348958 Supplier Novus Biologicals Supplier No. H00023339B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

VPS39 Polyclonal antibody specifically detects VPS39 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS39
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. NP_056104.2
Gene Alias hVam6p, KIAA0770FLJ21681, TLP, TRAP1-like protein, vacuolar protein sorting 39 (yeast), vacuolar protein sorting 39 homolog (S. cerevisiae), vam6/Vps39-like protein, VAM6FLJ46546
Host Species Mouse
Immunogen VPS39 (NP_056104.2, 1 a.a. - 875 a.a.) full-length human protein. MHDAFEPVPILEKLPLQIDCLAAWEEWLLVGTKQGHLLLYRIRKDVGCNRFEVTLEKSNKNFSKKIQQIHVVSQFKILVSLLENNIYVHDLLTFQQITTVSKAKGASLFTCDLQHTETGEEVLRMCVAVKKKLQLYFWKDREFHELQGDFSVPDVPKSMAWCENSICVGFKRDYYLIRVDGKGSIKELFPTGKQLEPLVAPLADGKVAVGQDDLTVVLNEEGICTQKCALNWTDIPVAMEHQPPYIIAVLPRYVEIRTFEPRLLVQSIELQRPRFITSGGSNIIYVASNHFVWRLIPVPMATQIQQLLQDKQFELALQLAEMKDDSDSEKQQQIHHIKNLYAFNLFCQKRFDESMQVFAKLGTDPTHVMGLYPDLLPTDYRKQLQYPNPLPVLSGAELEKAHLALIDYLTQKRSQLVKKLNDSDHQSSTSPLMEGTPTIKSKKKLLQIIDTTLLKCYLHTNVALVAPLLRLENNHCHIEESEHVLKKAHKYSELIILYEKKGLHEKALQVLVDQSKKANSPLKGHERTVQYLQHLGTENLHLIFSYSVWVLRDFPEDGLKIFTEDLPEVESLPRDRVLGFLIENFKGLAIPYLEHIIHVWEETGSRFHNCLIQLYCEKVQGLMKEYLLSFPAGKTPVPAGEEEGELGEYRQKLLMFLEISSYYDPGRLICDFPFDGLLEERALLLGRMGKHEQALFIYVHILKDTRMAEEYCHKHYDRNKDGNKDVYLSLLRMYLSPPSIHCLGPIKLELLEPKANLQAALQVLELHHSKLDTTKALNLLPANTQINDIRIFLEKVLEENAQKKRFNQVLKNLLHAEFLRVQEERILHQQVKCIITEEKVCMVCKKKIGNSAFARYPNGVVVHYFCSKEVNPADT
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23339
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.