missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
VPS39 Polyclonal antibody specifically detects VPS39 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | VPS39 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | hVam6p, KIAA0770FLJ21681, TLP, TRAP1-like protein, vacuolar protein sorting 39 (yeast), vacuolar protein sorting 39 homolog (S. cerevisiae), vam6/Vps39-like protein, VAM6FLJ46546 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of human VPS39. The immunogen is located within amino acids 710 - 760 of VPS39. Amino Acid Squence: HKHYDRNKDGNKDVYLS |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?