Learn More
VPS37C Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27128-1-AP-150UL
This item is not returnable.
View return policy
Description
The VPS37C antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human VPS37C. This antibody recognizes human antigen. The VPS37C antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| VPS37C | |
| 450 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| ESCRT I complex subunit VPS37C, hVps37C, PML39, VPS37C | |
| 150 μL | |
| 55048 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |