missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS37A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11870-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene belongs to the VPS37 family, and encodes a component of the ESCRT-I (endosomal sorting complex required for transport I) protein complex, required for the sorting of ubiquitinated transmembrane proteins into internal vesicles of multivesicular bodies. Expression of this gene is downregulated in hepatocellular carcinoma, and mutations in this gene are associated with autosomal recessive spastic paraplegia-53. A related pseudogene has been identified on chromosome 5. Alternatively spliced transcript variants have been found for this gene.Specifications
| VPS37A | |
| Polyclonal | |
| Unconjugated | |
| VPS37A | |
| ESCRT I complex subunit VPS37A, HCRP1, hVps37A, PQBP2, VPS37A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 137492, 52348 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8CHS8, Q8NEZ2 | |
| Vps37a | |
| VPS37A Fusion Protein Ag2453 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction