missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS37A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94426-0.02ml
This item is not returnable.
View return policy
Description
VPS37A Polyclonal antibody specifically detects VPS37A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| VPS37A | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| ESCRT-I complex subunit VPS37A, FLJ32642, HCRP1FLJ42616, hepatocellular carcinoma related protein 1, Hepatocellular carcinoma-related protein 1, hVps37A, polyglutamine binding protein 2, PQBP2, vacuolar protein sorting 37 homolog A (S. cerevisiae), vacuolar protein sorting 37A, vacuolar protein sorting 37A (yeast), vacuolar protein sorting-associated protein 37A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 224-397 of human VPS37A (NP_689628.2). MPDVPDAFPELSELSVSQLTDMNEQEEVLLEQFLTLPQLKQIITDKDDLVKSIEELARKNLLLEPSLEAKRQTVLDKYELLTQMKSTFEKKMQRQHELSESCSASALQARLKVAAHEAEEESDNIAEDFLEGKMEIDDFLSSFMEKRTICHCRRAKEEKLQQAIAMHSQFHAPL | |
| 0.02 mL | |
| Signal Transduction | |
| 137492 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction