missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPS35 Monoclonal Antibody (11H10), APC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545948
This item is not returnable.
View return policy
Description
A 1:1,000 dilution of MA5-45948 was sufficient for detection of VPS35 in 10 μg of SH-SY5Y by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary antibody. Detects approximately 92 kDa.
This gene belongs to a group of vacuolar protein sorting (VPS) genes. The encoded protein is a component of a large multimeric complex, termed the retromer complex, involved in retrograde transport of proteins from endosomes to the trans-Golgi network. The close structural similarity between the yeast and human proteins that make up this complex suggests a similarity in function. Expression studies in yeast and mammalian cells indicate that this protein interacts directly with VPS35, which serves as the core of the retromer complex.
Specifications
| VPS35 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| Q96QK1, Q9EQH3 | |
| VPS35 | |
| Full length recombinant human VSP35. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 11H10 | |
| APC | |
| VPS35 | |
| AI647796; DKFZp434E1211; DKFZp434P1672; FLJ10752; FLJ13588; FLJ20388; hVPS35; Maternal embryonic message 3; maternal-embryonic 3; Mem3; PARK17; TCCCTA00141; vacuolar protein sorting 35; vacuolar protein sorting 35 homolog; vacuolar protein sorting 35 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 35; Vesicle protein sorting 35; VPS35; VPS35 retromer complex component | |
| Mouse | |
| Protein G | |
| RUO | |
| 25479, 55737, 65114 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction