missing translation for 'onlineSavingsMsg'
Learn More

Vps34-IN-2, MedChemExpress

Product Code. 16403316
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16403316

Brand: Medchem Express HY12473

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vps34-IN-2 is a novel, potent and selective inhibitor of Vps34 with IC50s of 2 and 82 nM on the Vps34 enzymatic assay and the GFP-FYVE cellular assay, respectively. Vps34-IN-2 shows antiviral activity against SARS-CoV-2 (IC50 of 3.1 μM), HCoV-229E (IC50 of 0.7 μM) and HCoV-OC43.

Vps34-IN-2 is a novel, potent and selective inhibitor of Vps34 with IC50s of 2 and 82 nM on the Vps34 enzymatic assay and the GFP-FYVE cellular assay, respectively. Vps34-IN-2 shows antiviral activity against SARS-CoV-2 (IC50 of 3.1 μM), HCoV-229E (IC50 of 0.7 μM) and HCoV-OC43.

Specifications

Chemical Name or Material Vps34-IN-2
Physical Form Solid
CAS 1523404-29-6
Color Off-White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C18H25F3N4O3
For Use With (Application) Cancer-Kinase/protease
Solubility Information Ethanol : 50 mg/mL (124.25 mM; Need ultrasonic)
SMILES O=C1C=C(N2[C@H](C)COCC2)N=C3N(CC(C(C)C)=O)[C@H](C(F)(F)F)CCN31
Molecular Weight (g/mol) 402.41
Formula Weight 402.41
Percent Purity 99.49%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.