missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS34 (C terminal) Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12452-1-AP-150UL
This item is not returnable.
View return policy
Description
VPS34 (PIK3C3) is a catalytic subunit of the PI3K complex that mediates formation of phosphatidylinositol 3-phosphate; different complex forms are believed to play a role in multiple membrane trafficking pathways: PI3KC3-C1 is involved in initiation of autophagosomes and PI3KC3-C2 in maturation of autophagosomes and endocytosis. Involved in regulation of degradative endocytic trafficking and required for the abcission step in cytokinesis, probably in the context of PI3KC3-C2. Involved in the transport of lysosomal enzyme precursors to lysosomes. Required for transport from early to late endosomes.Specifications
| VPS34 (C terminal) | |
| Polyclonal | |
| Unconjugated | |
| PIK3C3 | |
| hVps34, PI 3 Kinase Class 3, PI 3-kinase p100, PI3K p100, PI3K type 3, PIK3C3, PtdIns 3 kinase type 3, VPS34 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 225326, 5289, 65052 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88763, Q6PF93, Q8NEB9 | |
| PIK3C3 | |
| VPS34 (C terminal) Fusion Protein Ag3110 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction